CPI-613 ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Devimistat, 6,8-Bis(benzylthio)octanoic acid
B5368-25MG
B5368-5MG
TCB5368-25MGEA
389
USD
TCB5368-25MG
TCB5368-5MG
CPI-613 ≥98.0% (by HPLC, titration analysis)
CPI-613
Formula:
C₂₂H₂₈O₂S₂ MW: 388.59 g/mol |
MDL Number:
MFCD22420826 CAS Number: 95809-78-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...