Triethyleneglycol divinyl ether ≥98.0% (by GC) stabilised

Supplier: TCI America

Synonyms: 3,6,9,12-Tetraoxatetradeca-1,13-diene

Stabilization: Stabilised with potassium hydroxide

T3460-100G T3460-25G
TCT3460-100GEA 251.84 USD
TCT3460-100G TCT3460-25G
Triethyleneglycol divinyl ether ≥98.0% (by GC) stabilised
Triethyleneglycol divinyl ether
Formula: C₁₀H₁₈O₄
MW: 202.2475 g/mol
Boiling Pt: 126°C
MDL Number: MFCD00074891
CAS Number: 765-12-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.0020 to 1.0060
Refractive index n20/D 1.4510 to 1.4550


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR