Ethylboronic acid pinacol ester ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethane, Ethylboronic acid Pinacol ester

E1116-1G E1116-5G
TCE1116-1GEA 50 USD
TCE1116-1G TCE1116-5G
Ethylboronic acid pinacol ester ≥95.0% (by GC, titration analysis)
Ethylboronic acid pinacol ester
Formula: C₈H₁₇BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19441153
CAS Number: 82954-89-0
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR