5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M2935-1G M2935-5G
TCM2935-1GEA 74 USD
TCM2935-1G TCM2935-5G
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid ≥98.0% (by HPLC, titration analysis)
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid
Formula: C₁₀H₉N₃O₂
MW: 203.197 g/mol
MDL Number: MFCD18382679
CAS Number: 956317-36-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR