AG-494 ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Danger
|
Synonyms:
Tyrphostin AG 494, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-phenylacrylamide
A2704-100MG
A2704-20MG
TCA2704-100MGEA
259.4
USD
TCA2704-100MG
TCA2704-20MG
AG-494 ≥98.0% (by HPLC, total nitrogen)
AG-494
Formula:
C₁₆H₁₂N₂O₃ MW: 280.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00209863 CAS Number: 133550-35-3 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...