Tris(pyrazol-1-yl)methane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Tris(1-pyrazolyl)methane

T3026-1G T3026-5G
TCT3026-1GEA 185 USD
TCT3026-1G TCT3026-5G
Tris(pyrazol-1-yl)methane ≥98.0% (by GC)
Tris(pyrazol-1-yl)methane
Formula: C₁₀H₁₀N₆
MW: 214.23 g/mol
MDL Number: MFCD00040255
CAS Number: 80510-03-8

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR