Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)

Supplier: TCI America

Synonyms: [NH2Me2][(RuCl((S)-xylbinap))2(μ-Cl)3]

N1020-1G N1020-200MG
TCN1020-1GEA 204 USD
TCN1020-1G TCN1020-200MG
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Formula: C₁₀₆H₁₀₄Cl₅NP₄Ru₂
MW: 1895.29 g/mol
MDL Number: MFCD09753025
CAS Number: 944451-10-9

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR