Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)

Supplier: TCI America

Synonyms: Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole]diruthenate(II), [NH2Me2][(RuCl((R)-dm-segphos®))2(mu-Cl)3], [NH2Me2][{RuCl((R)-dm-segphos®)}2(mu-Cl)3]

N1023-1G N1023-200MG
TCN1023-1GEA 191.4 USD
TCN1023-1G TCN1023-200MG
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Formula: C₉₄H₉₆Cl₅NO₈P₄Ru₂
MW: 1871.09 g/mol
MDL Number: MFCD09753038
CAS Number: 935449-46-0

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR