Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)

Supplier: TCI America
Danger

N1018-1G N1018-200MG
TCN1018-1GEA 150 USD
TCN1018-1G TCN1018-200MG
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Formula: C₉₈H₈₈Cl₅NP₄Ru₂
MDL Number: MFCD09753034
CAS Number: 309735-86-2

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR