Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)

Supplier: TCI America

N1016-1G N1016-200MG
TCN1016-1GEA 150.6 USD
TCN1016-1G TCN1016-200MG
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Formula: C₉₀H₇₂Cl₅NP₄Ru₂
MDL Number: MFCD09753033
CAS Number: 199541-17-8

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR