Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)

Supplier: TCI America
Danger

N1021-1G N1021-200MG
TCN1021-1GEA 152.7 USD
TCN1021-1G TCN1021-200MG
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Formula: C₇₈H₆₄Cl₅NO₈P₄Ru₂
MDL Number: MFCD09753037
CAS Number: 346457-41-8

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR