Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Supplier: TCI America
|
Danger
|
N1017-1G
N1017-200MG
TCN1017-1GEA
133.9
USD
TCN1017-1G
TCN1017-200MG
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Formula:
C₉₈H₈₈Cl₅NP₄Ru₂ |
MDL Number:
MFCD09753034 CAS Number: 749935-02-2 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...