4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Acetoxyphenylboronic acid pinacol ester, 2-(4-Acetoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl Acetate, 4-Acetoxyphenylboronic acid, pinacol ester, 4-Acetoxyphenylboronic acid Pinacol Ester

T3224-1G T3224-5G
TCT3224-1GEA 63 USD
TCT3224-1G TCT3224-5G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥96.0% (by GC, titration analysis)
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
Formula: C₁₄H₁₉BO₄
MDL Number: MFCD03453060
CAS Number: 480424-70-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR