2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: t-BuXPhos, Di-tert-butyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine, 2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, tBuXPhos

D5039-1G D5039-5G
TCD5039-1GEA 135 USD
TCD5039-1G TCD5039-5G
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl ≥98.0% (by HPLC, titration analysis)
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl
Formula: C₂₉H₄₅P
MW: 424.65 g/mol
MDL Number: MFCD06411306
CAS Number: 564483-19-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR