(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by GC)
Supplier: TCI America
B3203-1G
B3203-5G
TCB3203-1GEA
138.1
USD
TCB3203-1G
TCB3203-5G
(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by GC)
(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
Formula:
C₂₄H₂₂O₄ |
MDL Number:
MFCD03788935 CAS Number: 142128-92-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Specific rotation [a]20/D | -90.0 to -95.0 deg (C=1,CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...