(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (R)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl

D4882-1G D4882-5G
76219-740EA 160 USD
76219-740 76219-742
(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by HPLC, titration analysis)
(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
Formula: C₂₄H₂₀Br₂O₄
MW: 532.23 g/mol
MDL Number: MFCD03788936
CAS Number: 179866-74-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 133.0 to 137.0 deg-C
Specific rotation [a]20/D +20.0 to +23.0 deg (C=1,CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR