NSC 66811 ≥98.0% (by GC)

Supplier: TCI America

Synonyms: NSC66811, 2-Methyl-7-[phenyl(phenylamino)methyl]-8-quinolinol, 2-Methyl-7-(phenyl(phenylamino)methyl)quinolin-8-ol, 8-Hydroxy-2-methyl-7-[phenyl(phenylamino)methyl]quinoline

M2390-100MG M2390-25MG
TCM2390-100MGEA 173 USD
TCM2390-100MG TCM2390-25MG
NSC 66811 ≥98.0% (by GC)
NSC 66811
Formula: C₂₃H₂₀N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD08823776
CAS Number: 6964-62-1

Order Now

Specification Test Results

Appearance White to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR