GBR 12909 dihydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0541-250MG G0541-50MG
TCG0541-250MGEA 541 USD
TCG0541-250MG TCG0541-50MG
GBR 12909 dihydrochloride ≥98.0% (by titrimetric analysis)
GBR 12909 dihydrochloride

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 219.0 to 223.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR