(-)-Arctigenin ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Arctigenin, (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]dihydro-3-[(4-hydroxy-3-methoxyphenyl)methyl]-2(3H)-furanone, (3R,4R)-4-(3,4-Dimethoxybenzyl)-3-(4-hydroxy-3-methoxybenzyl)dihydrofuran-2(3H)-one

A3117-10MG A3117-50MG
76219-290EA 97 USD
76219-290 76219-292
(-)-Arctigenin ≥95.0% (by GC)
(-)-Arctigenin
Formula: C₂₁H₂₄O₆
MW: 372.4 g/mol
MDL Number: MFCD00870597
CAS Number: 7770-78-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Specific rotation [a]20/D -40.0 to -44.0 deg (C=1, methanol)
Purity (GC) min. 95.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR