SB-525334 ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 6-[2-(tert-Butyl)-5-(6-methylpyridin-2-yl)-1H-imidazol-4-yl]quinoxaline, 6-(2-(tert-Butyl)-5-(6-methylpyridin-2-yl)-1H-imidazol-4-yl)quinoxaline

B5776-10MG B5776-50MG
TCB5776-10MGEA 195 USD
TCB5776-10MG TCB5776-50MG
SB-525334 ≥98.0% (by HPLC)
SB-525334
Formula: C₂₁H₂₁N₅
Storage Temperature: Refrigerator
MDL Number: MFCD11045307
CAS Number: 356559-20-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 156.0 to 160.0 deg-C
Elemental analysis (Nitrogen) 19.40 to 21.40 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR