Flt-3 inhibitor ≥98.0% (by HPLC)

Supplier: TCI America

D5275-100MG D5275-25MG
76219-890EA 431 USD
76219-890 76219-892
Flt-3 inhibitor ≥98.0% (by HPLC)
Flt-3 inhibitor
Formula: C₁₈H₂₀N₂O₄S
Storage Temperature: Freezer
MDL Number: MFCD00617269
CAS Number: 301305-73-7

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.30 to 8.30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR