Lomitapide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-(2,2,2-Trifluoroethyl)-9-[4-[4-[4'-(trifluoromethyl)-[1,1'-biphenyl]-2-ylcarboxamido]piperidin-1-yl]butyl]-9H-fluorene-9-carboxamide, AEGR-733, BMS-201038

L0298-100MG L0298-25MG
TCL0298-100MGEA 303 USD
TCL0298-100MG TCL0298-25MG
Lomitapide ≥98.0% (by HPLC)
Lomitapide
Formula: C₃₉H₃₇F₆N₃O₂
MW: 693.73 g/mol
Melting Pt: 142°C
Storage Temperature: Refrigerator
MDL Number: MFCD16620494
CAS Number: 182431-12-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.75 to 6.35 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR