Roflumilast ≥98.0% (by HPLC)

Supplier: TCI America
Warning

R0193-1G R0193-250MG
TCR0193-1GEA 166 USD
TCR0193-1G TCR0193-250MG
Roflumilast ≥98.0% (by HPLC)
Roflumilast
Formula: C₁₇H₁₄Cl₂F₂N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00938270
CAS Number: 162401-32-3
Merck Index: 14,08249

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 157.0 to 161.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR