TDZD-8 ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione

B4436-25MG B4436-5MG
TCB4436-25MGEA 266 USD
TCB4436-25MG TCB4436-5MG
TDZD-8 ≥98.0% (by HPLC)
TDZD-8
Formula: C₁₀H₁₀N₂SO₂
MW: 222.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04973552
CAS Number: 327036-89-5

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 12.00 to 13.00 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR