5-Norbornene-2-methanol ≥98.0% (by GC)

Supplier: TCI America
Warning

N0979-25G N0979-5G
TCN0979-25GEA 210.2 USD
TCN0979-25G TCN0979-5G
5-Norbornene-2-methanol ≥98.0% (by GC)
5-Norbornene-2-methanol
Formula: C₈H₁₂O
Boiling Pt: 120°C
Flash Pt: 87°C
MDL Number: MFCD00167571
CAS Number: 95-12-5
EINECS: 202-392-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR