N-(2-Hydroxyethyl)maleimide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
Synonyms:
1-(2-Hydroxyethyl)-1H-pyrrole-2,5-dione, 1-(2-Hydroxyethyl)maleimide, N-(Ethanol)maleimide, 1-(2-Hydroxyethyl)-pyrrole-2,5-dione, HEMI
H1544-1G
H1544-5G
TCH1544-1GEA
190
USD
TCH1544-1G
TCH1544-5G
N-(2-Hydroxyethyl)maleimide ≥98.0% (by HPLC, total nitrogen)
N-(2-Hydroxyethyl)maleimide
Formula:
C₆H₇NO₃ MW: 141.13 g/mol Boiling Pt: 130°C |
MDL Number:
MFCD00465266 CAS Number: 1585-90-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...