Febuxostat ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(3-Cyano-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylic acid

F0847-1G F0847-5G
TCF0847-1GEA 101.37 USD
TCF0847-1G TCF0847-5G
Febuxostat ≥97.0% (by HPLC, titration analysis)
Febuxostat
Formula: C₁₆H₁₆N₂O₃S
MW: 316.37 g/mol
MDL Number: MFCD00871598
CAS Number: 144060-53-7
Merck Index: 14,03948

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 207.0 to 211.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR