Isothiazole ≥98.0% (by GC)

Supplier: TCI America
Danger

I0982-1G I0982-200MG
TCI0982-1GEA 148 USD
TCI0982-1G TCI0982-200MG
Isothiazole ≥98.0% (by GC)
Isothiazole
Formula: C₃H₃NS
Boiling Pt: 114°C
MDL Number: MFCD00020818
CAS Number: 288-16-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR