Fusidic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoic acid, Fusidine

F1007-1G F1007-5G
TCF1007-1GEA 216.6 USD
TCF1007-1G TCF1007-5G
Fusidic acid ≥97.0% (by HPLC, titration analysis)
Fusidic acid
Formula: C₃₁H₄₈O₆
Melting Pt: 193°C
Storage Temperature: Refrigerator
MDL Number: MFCD00865135
CAS Number: 6990-06-3
EINECS: 230-256-0
Merck Index: 14,04317

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -6.0 to -10.0 deg (C=1,CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR