Mal-amido-PEG4-acid ≥95.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic acid, 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic acid
M2666-100MG
M2666-25MG
TCM2666-100MGEA
161.1
USD
TCM2666-100MG
TCM2666-25MG
Mal-amido-PEG4-acid ≥95.0% (by HPLC)
Mal-amido-PEG4-acid
Formula:
C₁₈H₂₈N₂O₉ MW: 416.42 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13184954 CAS Number: 1263045-16-4 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 6.10 to 7.40 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...