Manganesebis(trifluoromethylsulfonyl)imide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Manganese(II) triflimide, Manganese(II) Bis(trifluoromethanesulphonyl)imide, Manganese bis(trifluoromethanesulphonyl)imide, Bis(trifluoromethanesulphonyl)imide manganese(II) salt

M2862-1G M2862-5G
TCM2862-1GEA 137 USD
TCM2862-1G TCM2862-5G
Manganesebis(trifluoromethylsulfonyl)imide ≥97.0% (by titrimetric analysis)
Manganesebis(trifluoromethylsulfonyl)imide
Formula: C₄F₁₂MnN₂O₈S₄
MW: 615.21 g/mol
MDL Number: MFCD23380173
CAS Number: 207861-55-0
UN: 1759
ADR: 8 II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR