N'-Hydroxypyridine-4-carboximidamide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
N-Hydroxyisonicotinimidamide, 4-Pyridylamidoxime, Pyridine-4-amidoxime, N'-Hydroxy-4-pyridinecarboximidamide
P2591-1G
P2591-5G
TCP2591-1GEA
33
USD
TCP2591-1G
TCP2591-5G
N'-Hydroxypyridine-4-carboximidamide ≥98.0% (by HPLC, titration analysis)
N'-Hydroxypyridine-4-carboximidamide
Formula:
C₆H₇N₃O MW: 137.142 g/mol |
MDL Number:
MFCD00125873 CAS Number: 1594-57-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 203.0 to 207.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...