2-Ethylhexyl palmitate ≥98.0% (by GC)

Supplier: TCI America

E1435-250G E1435-25G
TCE1435-250GEA 122 USD
TCE1435-250G TCE1435-25G
2-Ethylhexyl palmitate ≥98.0% (by GC)
2-Ethylhexyl palmitate
Formula: C₂₄H₄₈O₂
MDL Number: MFCD00072255
CAS Number: 29806-73-3
EINECS: 249-862-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.8550 to 0.8650


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR