Magnesiumbis(trifluoromethylsulfonyl)imide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
3-nitro-1H-pyrimidine-2,4-dione, Magnesium(II) triflimide, Bis(trifluoromethanesulphonyl)imide Magnesium(II) salt, 2,4(1H,3H)-Pyrimidinedione, nitro-, Magnesium bis(trifluoromethylsulphonyl)imide, Magnesium(II) Bis(trifluoromethanesulphonyl)imide, 3-Nitropyrimidine-2,4(1H,3H)-dione
M2861-1G
M2861-5G
TCM2861-1GEA
124
USD
TCM2861-1G
TCM2861-5G
Magnesiumbis(trifluoromethylsulfonyl)imide ≥97.0% (by titrimetric analysis)
Magnesiumbis(trifluoromethylsulfonyl)imide
Formula:
C₄F₁₂MgN₂O₈S₄ MW: 584.6 g/mol |
MDL Number:
MFCD14156020 CAS Number: 133395-16-1 UN: 1759 ADR: 8 II |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Purity (Ion exchange titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...