4-(Methanesulfonamido)phenylboronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-[4-(Methanesulphonylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-(Methylsulphonyl)-4-(4,4,5,5-tetraMethyl-1,3,2-dioxaborolan-2-yl)benzenamine, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulphonamide, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenyl)methanesulphonamide, 4-(Methanesulphonylamino)phenylboronic acid Pinacol ester, 4-Methanesulphonylaminophenylboronic acid, pinacol ester, N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulphonamide, 4-(Methylsulphonamido)phenylboronic acid pinacol ester

T3557-1G T3557-5G
TCT3557-1GEA 56 USD
TCT3557-1G TCT3557-5G
4-(Methanesulfonamido)phenylboronic acid pinacol ester ≥98.0% (by titrimetric analysis)
4-(Methanesulfonamido)phenylboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₄S
MW: 297.18 g/mol
MDL Number: MFCD05663876
CAS Number: 616880-14-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR