PD-0173074 ≥95.0% (by HPLC)
Supplier: TCI America
|
Warning
|
P2474-10MG
P2474-50MG
TCP2474-10MGEA
126
USD
TCP2474-10MG
TCP2474-50MG
PD-0173074 ≥95.0% (by HPLC)
PD-0173074
Formula:
C₂₈H₄₁N₇O₃ Melting Pt: 86°C Storage Temperature: Refrigerator |
MDL Number:
MFCD08705327 CAS Number: 219580-11-7 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 18.00 to 19.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...