N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan) ≥98.0% (by GC)

Supplier: TCI America

D4678-1G D4678-200MG
76219-054EA 213 USD
76219-054 76219-056
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan) ≥98.0% (by GC)
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan)
Formula: C₁₅H₁₇NO
MDL Number: MFCD00056615
CAS Number: 70504-01-7

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR