N-Methoxy-N-methylbenzamide ≥98.0% (by GC)

Supplier: TCI America
Warning

M2672-25G M2672-5G
TCM2672-25GEA 254 USD
TCM2672-25G TCM2672-5G
N-Methoxy-N-methylbenzamide ≥98.0% (by GC)
N-Methoxy-N-methylbenzamide
Formula: C₉H₁₁NO₂
Boiling Pt: 112°C
MDL Number: MFCD00075320
CAS Number: 6919-61-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.1050 to 1.1090
Refractive index n20/D 1.5320 to 1.5360


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR