Flumazenil ≥99.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate

F0958-100MG F0958-25MG
TCF0958-100MGEA 259 USD
TCF0958-100MG TCF0958-25MG
Flumazenil ≥99.0% (by GC)
Flumazenil
Formula: C₁₅H₁₄FN₃O₃
MW: 303.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00242764
CAS Number: 78755-81-4
Merck Index: 14,04135

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR