Ethyl-4-(1-piperazinyl)benzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 4-(piperazin-1-yl)benzoate

E1260-25G E1260-5G
TCE1260-25GEA 393 USD
TCE1260-25G TCE1260-5G
Ethyl-4-(1-piperazinyl)benzoate ≥98.0% (by GC)
Ethyl-4-(1-piperazinyl)benzoate
Formula: C₁₃H₁₈N₂O₂
MW: 234.30 g/mol
MDL Number: MFCD04973340
CAS Number: 80518-57-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR