Carbonylhydridotris(triphenylphosphine)iridium(I)

Supplier: TCI America
Danger

Synonyms: Hydridocarbonyltris(triphenylphosphine)iridium(I)

C3040-1G C3040-200MG
TCC3040-1GEA 743 USD
TCC3040-1G TCC3040-200MG
Carbonylhydridotris(triphenylphosphine)iridium(I)
Carbonylhydridotris(triphenylphosphine)iridium(I)
Formula: C₅₅H₄₆IrOP₃
MW: 1008.12 g/mol
MDL Number: MFCD00015525
CAS Number: 17250-25-8
EINECS: 241-282-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Elemental analysis (Carbon) 62.00 to 69.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR