Bisindolylmaleimide I ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 3-(1-(3-(Dimethylamino)propyl)-1H-indol-3-yl)-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3-[1-[3-(Dimethylamino)propyl]-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, GF 109203X

B5781-25MG B5781-5MG
TCB5781-25MGEA 298 USD
TCB5781-25MG TCB5781-5MG
Bisindolylmaleimide I ≥97.0% (by HPLC)
Bisindolylmaleimide I
Formula: C₂₅H₂₄N₄O₂
MW: 412.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236428
CAS Number: 133052-90-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 268.0 to 272.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR