Anthracene-9,10-diboronic acid bis(pinacol) ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 9,10-Anthracenediboronic acid bis(pinacol) ester, Anthracene-9,10-diboronic acid bis(pinacol) ester, 9,10-Bis(4,4,5,5-tetramethyl[1.3.2]dioxaborolan-2-yl)anthracene, 9,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene

B4562-25G B4562-5G
TCB4562-25GEA 1268 USD
TCB4562-25G TCB4562-5G
Anthracene-9,10-diboronic acid bis(pinacol) ester ≥98.0% (by GC)
Anthracene-9,10-diboronic acid bis(pinacol) ester
Formula: C₂₆H₃₂B₂O₄
MDL Number: MFCD16294538
CAS Number: 863992-56-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR