7,8-Dihydroxy-4-methylcoumarin ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Methyldaphnetin, 7,8-Dihydroxy-4-methyl-2H-chromen-2-one
D4793-1G
D4793-5G
TCD4793-1GEA
131
USD
TCD4793-1G
TCD4793-5G
7,8-Dihydroxy-4-methylcoumarin ≥98.0% (by GC, titration analysis)
7,8-Dihydroxy-4-methylcoumarin
Formula:
C₁₀H₈O₄ MW: 192.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00016972 CAS Number: 2107-77-9 EINECS: 218-290-4 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 241.0 to 245.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...