5-Bromo-2-phenylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5018-1G B5018-5G
TCB5018-1GEA 230 USD
TCB5018-1G TCB5018-5G
5-Bromo-2-phenylpyridine ≥98.0% (by GC)
5-Bromo-2-phenylpyridine
Formula: C₁₁H₈BrN
MDL Number: MFCD04114181
CAS Number: 27012-25-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR