4-Ethoxy-2-methylbenzeneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Ethoxy-2-methylphenylboronic acid, 4-Ethoxy-2-methylbenzeneboronic acid, B-(4-Ethoxy-2-methylphenyl)boronic acid
E1173-1G
E1173-5G
TCE1173-1GEA
47
USD
TCE1173-1G
TCE1173-5G
4-Ethoxy-2-methylbenzeneboronic acid (contains varying amounts of Anhydride)
4-Ethoxy-2-methylbenzeneboronic acid
Formula:
C₉H₁₃BO₃ |
MDL Number:
MFCD04115659 CAS Number: 313545-31-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...