4-(Benzyloxycarbonyl)benzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (4-((Benzyloxy)carbonyl)phenyl)boronic acid, 4-Benzyloxycarbonylphenylboronic acid, 4-Carbobenzoxybenzeneboronic acid, 4-(Benzyloxycarbonyl)benzeneboronic acid, 4-Carbobenzoxyphenylboronic acid, 4-(Benzyloxycarbonyl)phenylboronic acid

B4725-1G B4725-5G
TCB4725-1GEA 137 USD
TCB4725-1G TCB4725-5G
4-(Benzyloxycarbonyl)benzeneboronic acid (contains varying amounts of Anhydride)
4-(Benzyloxycarbonyl)benzeneboronic acid
Formula: C₁₄H₁₃BO₄
MW: 256.06 g/mol
MDL Number: MFCD02179442
CAS Number: 184000-11-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR