4-(6-Amino-3-pyridyl)-1-boc-piperazine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Boc-4-(6-amino-3-pyridyl)piperazine, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate, tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate, 1-tert-Butoxycarbonyl-4-(6-amino-3-pyridyl)piperazine, 4-(6-Amino-3-pyridyl)piperazine-1-carboxylic acid tert-Butyl ester
B4669-1G
B4669-200MG
TCB4669-1GEA
248
USD
TCB4669-1G
TCB4669-200MG
4-(6-Amino-3-pyridyl)-1-boc-piperazine ≥98.0% (by HPLC, titration analysis)
4-(6-Amino-3-pyridyl)-1-boc-piperazine
Formula:
C₁₄H₂₂N₂O₄ |
MDL Number:
MFCD11594962 CAS Number: 571188-59-5 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...