3-Ethoxy-5-fluorobenzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

E1205-1G E1205-200MG
TCE1205-1GEA 231 USD
TCE1205-1G TCE1205-200MG
3-Ethoxy-5-fluorobenzeneboronic acid (contains varying amounts of Anhydride)
3-Ethoxy-5-fluorobenzeneboronic acid
Formula: C₈H₁₀BFO₃
MDL Number: MFCD07363779
CAS Number: 850589-53-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR