3-(2-Naphthyl)benzeneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
3-(2-Naphthyl)benzeneboronic acid, (3-(Naphthalen-2-yl)phenyl)boronic acid, 3-(2-Naphthyl)phenylboronic acid
N1009-1G
N1009-5G
TCN1009-1GEA
80.6
USD
TCN1009-1G
TCN1009-5G
3-(2-Naphthyl)benzeneboronic acid (contains varying amounts of Anhydride)
3-(2-Naphthyl)benzeneboronic acid
Formula:
C₁₆H₁₃BO₂ MW: 248.09 g/mol |
MDL Number:
MFCD16293738 CAS Number: 870774-29-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...